4-(Ethoxycarbonyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid
OR10470-100G
OR10470-25G
OR10470-5G
APOSOR10470-100GEA
148
EUR
APOSOR10470-100G
APOSOR10470-25G
APOSOR10470-5G
4-(Ethoxycarbonyl)phenylboronsäure
4-(Ethoxycarbonyl)phenylboronsäure
Formel:
C₉H₁₁BO₄ Schmelzpunkt: 135…140 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179441 CAS-Nummer: 4334-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...