4-(Ethoxycarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Ethoxycarbonyl)benzeneboronic acid, (4-Ethoxycarbonylphenyl)boronic acid

OR10470-100G OR10470-25G OR10470-5G
APOSOR10470-100GEA 148 EUR
APOSOR10470-100G APOSOR10470-25G APOSOR10470-5G
4-(Ethoxycarbonyl)phenylboronsäure
4-(Ethoxycarbonyl)phenylboronsäure
Formel: C₉H₁₁BO₄
Molecular Weight: 193.99 g/mol
Schmelzpunkt: 135…140 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD02179441
CAS-Nummer: 4334-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR