5-Amino-3-brom-2-chlorpyridin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Brom-2-chlor-5-pyridylamin
OR1048-5G
APOSOR1048-5GEA
21.9
EUR
APOSOR1048-5G
5-Amino-3-brom-2-chlorpyridin
5-Amino-3-brom-2-chlorpyridin
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Schmelzpunkt: 118…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840433 CAS-Nummer: 130284-53-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...