4-(2-Aminoethyl)tetrahydropyran Hydrochlorid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Tetrahydro-2H-pyran-4-yl)ethanamine hydrochloride, 4-(2-Aminoethyl)tetrahydropyran, HCl
OR10492-1G
APOSOR10492-1GEA
95.7
EUR
APOSOR10492-1G
4-(2-Aminoethyl)tetrahydropyran Hydrochlorid 98%
4-(2-Aminoethyl)tetrahydropyran Hydrochlorid
Formel:
C₇H₁₆ClNO Molecular Weight: 165,66 g/mol Schmelzpunkt: 132…136 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411927 CAS-Nummer: 389621-77-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...