trans-Heptenylboronic acid 97%

Lieferant: Apollo Scientific
Achtung

OR10493-1G OR10493-5G
APOSOR10493-1GEA 0 EUR
APOSOR10493-1G APOSOR10493-5G
trans-Heptenylboronic acid 97%
trans-Heptenylboronic acid
Formel: C₇H₁₅BO₂
Schmelzpunkt: 107…111 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01074600
CAS-Nummer: 57404-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR