trans-1-Hexen-1-ylboronic acid 98%

Lieferant: Apollo Scientific
Achtung

OR10494-250MG
APOSOR10494-250MGEA 29 EUR
APOSOR10494-250MG
trans-1-Hexen-1-ylboronic acid 98%
trans-1-Hexen-1-ylboronic acid
Formel: C₆H₁₃BO₂
Schmelzpunkt: 62…63 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01074553
CAS-Nummer: 42599-18-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR