4-Hydroxy-2-methylphenylboronic acid

Lieferant: Apollo Scientific
Achtung

OR10497-1G
APOSOR10497-1GEA 34.8 EUR
APOSOR10497-1G
4-Hydroxy-2-methylphenylboronic acid
4-Hydroxy-2-methylphenylboronic acid
Formel: C₇H₉BO₃
Molecular Weight: 151.96 g/mol
Schmelzpunkt: 100…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788424
CAS-Nummer: 493035-82-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR