(1-Methyl-1H-imidazol-5-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Hydroxymethyl)-1-methyl-1H-imidazole
OR10499-5G
APOSOR10499-5GEA
149
EUR
APOSOR10499-5G
(1-Methyl-1H-imidazol-5-yl)methanol
(1-Methyl-1H-imidazol-5-yl)methanol
Formel:
C₅H₈N₂O Molecular Weight: 112.1298 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179522 CAS-Nummer: 38993-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...