4-(Hydroxymethyl)phenylboronsäure 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR10501-100G
OR10501-25G
OR10501-5G
APOSOR10501-100GEA
157
EUR
APOSOR10501-100G
APOSOR10501-25G
APOSOR10501-5G
4-(Hydroxymethyl)phenylboronsäure 97+%
4-(Hydroxymethyl)phenylboronsäure
Formel:
C₇H₉BO₃ Schmelzpunkt: 251…256 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00792672 CAS-Nummer: 59016-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...