2-Hydroxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR10503-25G
OR10503-5G
OR10503-100G
APOSOR10503-25GEA
59.6
EUR
APOSOR10503-25G
APOSOR10503-5G
APOSOR10503-100G
2-Hydroxyphenylboronsäure
2-Hydroxyphenylboronsäure
Formel:
C₆H₇BO₃ Schmelzpunkt: 185 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01074581 CAS-Nummer: 89466-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...