4-(Hydroxymethyl)indol
Lieferant: Apollo Scientific
|
Achtung
|
OR10506-1G
APOSOR10506-1GEA
0
EUR
APOSOR10506-1G
4-(Hydroxymethyl)indol
4-(Hydroxymethyl)indole
Formel:
C₉H₉NO Schmelzpunkt: 46…50 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01632220 CAS-Nummer: 1074-85-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...