7-(Hydroxymethyl)indol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1H-Indol-7-yl)methanol

OR10509-500MG
APOSOR10509-500MGEA 174 EUR
APOSOR10509-500MG
7-(Hydroxymethyl)indol 97%
7-(Hydroxymethyl)indole
Formel: C₉H₉NO
Molecular Weight: 147,1739 g/mol
Schmelzpunkt: 44…48 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179597
CAS-Nummer: 1074-87-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR