4-(Isopropylcarbamoyl)phenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid
OR10521-1G
OR10521-5G
APOSOR10521-1GEA
75.4
EUR
APOSOR10521-1G
APOSOR10521-5G
4-(Isopropylcarbamoyl)phenylboronsäure
4-(Isopropylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₄BNO₃ Schmelzpunkt: 162…167 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04039353 CAS-Nummer: 397843-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...