4-(Isopropylcarbamoyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid

OR10521-1G OR10521-5G
APOSOR10521-1GEA 75.4 EUR
APOSOR10521-1G APOSOR10521-5G
4-(Isopropylcarbamoyl)phenylboronsäure
4-(Isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₄BNO₃
Schmelzpunkt: 162…167 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04039353
CAS-Nummer: 397843-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR