4-Isopropylthiophenylboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Isopropylthio)benzeneboronic acid, 4-(Isopropylthio)phenylboronic acid
OR10525-250MG
APOSOR10525-250MGEA
29
EUR
APOSOR10525-250MG
4-Isopropylthiophenylboronic acid
4-Isopropylthiophenylboronic acid
Formel:
C₉H₁₃BO₂S Molecular Weight: 196,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411947 CAS-Nummer: 380427-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...