3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, 3-Methoxycarbonyl-5-nitrophenylboronic acid, 3-(Methoxycarbonyl)-5-nitrophenylboronic acid, (3-(Methoxycarbonyl)-5-nitrophenyl)boronic acid
OR10532-25G
APOSOR10532-25GEA
156
EUR
APOSOR10532-25G
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
3-Methoxycarbonyl-5-nitrobenzeneboronic acid
Formel:
C₈H₈BNO₆ Molecular Weight: 224.96 g/mol Schmelzpunkt: 250…255 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179469 CAS-Nummer: 117342-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...