2-(Methoxycarbonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

OR10535-1G OR10535-250MG OR10535-5G
APOSOR10535-1GEA 24.8 EUR
APOSOR10535-1G APOSOR10535-250MG APOSOR10535-5G
2-(Methoxycarbonyl)phenylboronsäure
2-(Methoxycarbonyl)phenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
Schmelzpunkt: 90…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179452
CAS-Nummer: 374538-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR