4-(Methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

OR10537-100G
APOSOR10537-100GEA 184 EUR
APOSOR10537-100G
4-(Methoxycarbonyl)phenylboronsäurepinakolester
4-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Molecular Weight: 262.11 g/mol
Schmelzpunkt: 77…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179438
CAS-Nummer: 171364-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR