Methyl-2-amino-1,3-thiazol-5-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-aminothiazole-5-carboxylate, 2-Aminothiazole-5-carboxylic acid methyll ester

OR10538-5G
APOSOR10538-5GEA 43.5 EUR
APOSOR10538-5G
Methyl-2-amino-1,3-thiazol-5-carboxylat
Methyl-2-aminothiazole-5-carboxylate
Formel: C₅H₆N₂O₂S
Molecular Weight: 158.1800 g/mol
Schmelzpunkt: 191 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788562
CAS-Nummer: 6633-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR