Methyl-2-brom-1,3-thiazol-5-carboxylat 99+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-bromothiazole-5-carboxylate
OR10539-25G
APOSOR10539-25GEA
62.5
EUR
APOSOR10539-25G
Methyl-2-brom-1,3-thiazol-5-carboxylat 99+%
Methyl-2-bromothiazole-5-carboxylate
Formel:
C₅H₄BrNO₂S Schmelzpunkt: 62,1…62.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03788565 CAS-Nummer: 54045-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...