4-Methyl-1-naphthalinboronsäure

Lieferant: Apollo Scientific
Achtung

OR10547-25G
APOSOR10547-25GEA 125 EUR
APOSOR10547-25G
4-Methyl-1-naphthalinboronsäure
4-Methyl-1-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₂
Molecular Weight: 186.02 g/mol
Schmelzpunkt: 229…235 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01632204
CAS-Nummer: 103986-53-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR