Methyloxindol-7-carboxylat 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-oxoindoline-7-carboxylate
OR10551-50MG
APOSOR10551-50MGEA
21.9
EUR
APOSOR10551-50MG
Methyloxindol-7-carboxylat 96%
Methyloxindol-7-carboxylat
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,19 g/mol Schmelzpunkt: 182…184 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179610 CAS-Nummer: 380427-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...