2-Tolylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methylbenzolboronsäure, 2-Methylphenylboronsäure, (2-Methylphenyl)metaborsäure, Dihydroxy(2-methylphenyl)boran
OR10556-25G
APOSOR10556-25GEA
21.9
EUR
APOSOR10556-25G
2-Tolylboronsäure
2-Tolylboronsäure
Formel:
C₇H₉BO₂ Molecular Weight: 135,96 g/mol Schmelzpunkt: 162…164 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00093526 CAS-Nummer: 16419-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...