(2-Methylsulphinylphenyl)boronic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR10558-100MG
APOSOR10558-100MGEA
72.5
EUR
APOSOR10558-100MG
(2-Methylsulphinylphenyl)boronic acid 95%
(2-Methylsulphinylphenyl)boronic acid
Formel:
C₇H₉BO₃S Molecular Weight: 184,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095261 CAS-Nummer: 850567-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...