3-(Methylsulphonylamino)phenylboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methylsulphonylaminophenylboronic acid, 3-(Methylsulphonylamino)phenylboronic Acid
OR10561-1G
APOSOR10561-1GEA
21.9
EUR
APOSOR10561-1G
3-(Methylsulphonylamino)phenylboronic acid 98%
3-(Methylsulphonylamino)phenylboronic acid
Formel:
C₇H₁₀BNO₄S Molecular Weight: 215.04 g/mol Schmelzpunkt: 90…96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179478 CAS-Nummer: 148355-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...