4-(Methylsulphonylamino)benzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methylsulphonylamino)phenylboronic acid, 4-Methylsulphonylaminophenylboronic acid, 4-(Methanesulphonamido)phenylboronic acid

OR10562-1G
APOSOR10562-1GEA 33.5 EUR
APOSOR10562-1G
4-(Methylsulphonylamino)benzeneboronic acid 95%
4-(Methylsulphonylamino)benzeneboronic acid
Formel: C₇H₁₀BNO₄S
Molecular Weight: 215,03 g/mol
Schmelzpunkt: 148…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179473
CAS-Nummer: 380430-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR