[(2-Methylsulphonyl)aminophenyl]boronic acid, pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanesulphonamide, 2-Methylsulphonylaminophenylboronic acid, pinacol ester
OR10563-1G
APOSOR10563-1GEA
33.5
EUR
APOSOR10563-1G
[(2-Methylsulphonyl)aminophenyl]boronic acid, pinacol ester
[(2-Methylsulphonyl)aminophenyl]boronic acid, pinacol ester
Formel:
C₁₃H₂₀BNO₄S Molecular Weight: 297,18 g/mol Schmelzpunkt: 130…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179479 CAS-Nummer: 380430-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...