2-(Methylsulfonyl)phenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methylsulphonyl)benzeneboronic acid, (2-(Methylsulphonyl)phenyl)boronic acid

OR10564-250MG
APOSOR10564-250MGEA 21.9 EUR
APOSOR10564-250MG
2-(Methylsulfonyl)phenylboronsäure 97%
2-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200,02 g/mol
Schmelzpunkt: 78…84 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179472
CAS-Nummer: 330804-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR