3-(Methylsulfonyl)phenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR10565-1G
OR10565-5G
APOSOR10565-1GEA
0
EUR
APOSOR10565-1G
APOSOR10565-5G
3-(Methylsulfonyl)phenylboronsäure 98%
3-(Methylsulfonyl)phenylboronsäure
Formel:
C₇H₉BO₄S Schmelzpunkt: 102…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092935 CAS-Nummer: 373384-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...