3-Nitrophenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR10571-5G
APOSOR10571-5GEA
21.9
EUR
APOSOR10571-5G
3-Nitrophenylboronsäure 97%
3-Nitrophenylboronsäure
Formel:
C₆H₆BNO₄ Schmelzpunkt: 284…285 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007193 CAS-Nummer: 13331-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...