4-Nitrophenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR10572-1G
OR10572-5G
OR10572-25G
APOSOR10572-1GEA
21.9
EUR
APOSOR10572-1G
APOSOR10572-5G
APOSOR10572-25G
4-Nitrophenylboronsäure
4-Nitrophenylboronsäure
Formel:
C₆H₆BNO₄ Schmelzpunkt: 285…290 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00161360 CAS-Nummer: 24067-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...