2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Phthalimidomethyl)benzeneboronic acid, pinacol ester, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)isoindoline-1,3-dione
OR10581-250MG
APOSOR10581-250MGEA
21.9
EUR
APOSOR10581-250MG
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 98%
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione
Formel:
C₂₁H₂₂BNO₄ Molecular Weight: 363,22 g/mol Schmelzpunkt: 166…171 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02179489 CAS-Nummer: 138500-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...