Tetrahydropyran-4-essigsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tetrahydropyranyl-4-acetic acid
OR10586-1G
OR10586-5G
OR10586-10G
APOSOR10586-1GEA
21.9
EUR
APOSOR10586-1G
APOSOR10586-5G
APOSOR10586-10G
Tetrahydropyran-4-essigsäure 97%
Tetrahydropyran-4-essigsäure
Formel:
C₇H₁₂O₃ Schmelzpunkt: 60…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631204 CAS-Nummer: 85064-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...