Tetrahydropyran-4-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrahydropyran-4-yl-carboxylic acid

OR10587-25G OR10587-5G OR10587-100G
APOSOR10587-25GEA 31.9 EUR
APOSOR10587-25G APOSOR10587-5G APOSOR10587-100G
Tetrahydropyran-4-carbonsäure 98%
Tetrahydropyran-4-carbonsäure
Formel: C₆H₁₀O₃
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031016
CAS-Nummer: 5337-03-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR