ɑ-[2-(Methylamino)ethyl]benzyl alcohol 98%

Lieferant: Apollo Scientific

Synonyme: 3-Hydroxy-N-methyl-3-phenylpropylamine, 3-(Methylamino)-1-phenylpropanol, α-[2-(Methylamino)ethyl]benzyl Alcohol, 3-(Methylamino)-1-phenylpropan-1-ol

OR1059-500G
APOSOR1059-500GEA 168 EUR
APOSOR1059-500G
ɑ-[2-(Methylamino)ethyl]benzyl alcohol 98%
alpha-[2-(Methylamino)ethyl]benzyl alcohol
Formel: C₁₀H₁₅NO
Molecular Weight: 165.24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00674078
CAS-Nummer: 42142-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR