2,3,6-Trimethoxyphenylboronsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,6-Trimethoxybenzeneboronic acid, (2,3,6-Trimethoxyphenyl)boronic acid

OR10594-250MG
APOSOR10594-250MGEA 21.9 EUR
APOSOR10594-250MG
2,3,6-Trimethoxyphenylboronsäure 95%
2,3,6-Trimethoxyphenylboronsäure
Formel: C₉H₁₃BO₅
Molecular Weight: 212,01 g/mol
Schmelzpunkt: 34…40 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03411944
CAS-Nummer: 380430-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR