2-Vinylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Dihydroxy(2-vinylphenyl)borane, (2-Vinylphenyl)metaboric acid, 2-Vinylbenzeneboronic acid

OR10598-250MG
APOSOR10598-250MGEA 21.9 EUR
APOSOR10598-250MG
2-Vinylphenylboronsäure
2-Vinylphenylboronsäure
Formel: C₈H₉BO₂
Molecular Weight: 147,97 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01074659
CAS-Nummer: 15016-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR