2-Amino-4-chloro-6-isopropylpyrimidine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-6-isopropylpyrimidin-2-amine

OR10626-250MG
APOSOR10626-250MGEA 21.9 EUR
APOSOR10626-250MG
2-Amino-4-chloro-6-isopropylpyrimidine
2-Amino-4-chloro-6-isopropylpyrimidine
Formel: C₇H₁₀ClN₃
Molecular Weight: 171,63 g/mol
Schmelzpunkt: 97…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840198
CAS-Nummer: 73576-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR