4,5-Dichlorphthalonitril 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR1063-250MG
OR1063-5G
OR1063-1G
APOSOR1063-250MGEA
40.6
EUR
APOSOR1063-250MG
APOSOR1063-5G
APOSOR1063-1G
4,5-Dichlorphthalonitril 95%
4,5-Dichlorphthalonitril
Formel:
C₈H₂Cl₂N₂ Schmelzpunkt: 180…184 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191408 CAS-Nummer: 139152-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...