1-Chloro-3-(triphenylphosphoranylidene)acetone

Lieferant: Apollo Scientific
Achtung

OR10637-250MG
APOSOR10637-250MGEA 0 EUR
APOSOR10637-250MG
1-Chloro-3-(triphenylphosphoranylidene)acetone
1-Chloro-3-(triphenylphosphoranylidene)acetone
Formel: C₂₁H₁₈ClOP
Lagertemperatur: Raumtemperatur
MDL: MFCD04038921
CAS-Nummer: 13605-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR