[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide 97%

Lieferant: Apollo Scientific
Achtung

OR10642-5G
APOSOR10642-5GEA 109 EUR
APOSOR10642-5G
[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide 97%
[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide
Formel: C₂₅H₂₈O₂P·Br
Molecular Weight: 471,366 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051866
CAS-Nummer: 118026-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR