(1-Ethoxycarbonylethyliden)triphenylphosphoran 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate
OR10643-100G
APOSOR10643-100GEA
78.3
EUR
APOSOR10643-100G
(1-Ethoxycarbonylethyliden)triphenylphosphoran 95%
(1-Ethoxycarbonylethyliden)triphenylphosphoran
Formel:
C₂₃H₂₃O₂P Molecular Weight: 362,41 g/mol Schmelzpunkt: 157…159 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00009160 CAS-Nummer: 5717-37-3 EINECS: 227-215-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...