(Triphenylphosphoranyliden)acetaldehyd 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane

OR10653-25G OR10653-100G
APOSOR10653-25GEA 21.9 EUR
APOSOR10653-25G APOSOR10653-100G
(Triphenylphosphoranyliden)acetaldehyd 95%
(Triphenylphosphoranylidene)acetaldehyde
Formel: C₂₀H₁₇OP
Molecular Weight: 304.33 g/mol
Schmelzpunkt: 185…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006994
CAS-Nummer: 2136-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR