Diethyl (3-hydroxyphenyl)phosphonate 98%

Lieferant: Apollo Scientific

OR10716-1G OR10716-500MG
APOSOR10716-1GEA 693 EUR
APOSOR10716-1G APOSOR10716-500MG
Diethyl (3-hydroxyphenyl)phosphonate 98%
Diethyl (3-hydroxyphenyl)phosphonate
Formel: C₁₀H₁₅O₄P
Lagertemperatur: Raumtemperatur
MDL: MFCD01112119
CAS-Nummer: 33733-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR