(3-Methylbenzyl)phosphonsäure
Lieferant: Apollo Scientific
|
Gefahr
|
OR10786-1G
OR10786-5G
APOSOR10786-1GEA
116
EUR
APOSOR10786-1G
APOSOR10786-5G
(3-Methylbenzyl)phosphonsäure
(3-Methylbenzyl)phosphonsäure
Formel:
C₈H₁₁O₃P Schmelzpunkt: 194…195 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
18945-65-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...