4-Chlor-2-methylanisol

Lieferant: Apollo Scientific

Synonyme: 5-Chlor-2-methoxytoluol, 1-Chlor-4-methoxy-3-methylbenzol

OR1079-25G
APOSOR1079-25GEA 104 EUR
APOSOR1079-25G
4-Chlor-2-methylanisol
4-Chlor-2-methylanisol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070771
CAS-Nummer: 3260-85-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR