Tetraethyl-p-xylylenediphosphonate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: p-Xylylenediphosphonic Acid Tetraethyl Ester, p-Bis(diethylphosphono)xylene

OR10800-5G OR10800-100G OR10800-25G
APOSOR10800-5GEA 108 EUR
APOSOR10800-5G APOSOR10800-100G APOSOR10800-25G
Tetraethyl-p-xylylenediphosphonate 98%
Tetraethyl-p-xylylenediphosphonate
Formel: C₁₆H₂₈O₆P₂
Siedepunkt: 220 °C/7mmHg
Schmelzpunkt: 78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00671576
CAS-Nummer: 4546-04-7
EINECS: 224-902-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR