Ethoxydiphenylphosphine 98%
Lieferant: Apollo Scientific
Synonyme:
Diphenylphosphinous Acid Ethyl Ester
OR10812-100G
APOSOR10812-100GEA
109
EUR
APOSOR10812-100G
Ethoxydiphenylphosphine 98%
Ethoxydiphenylphosphine
Formel:
C₁₄H₁₅OP Molecular Weight: 230,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009087 CAS-Nummer: 719-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...