3-Acetyl-1-(phenylsulphonyl)pyrrole

Lieferant: Apollo Scientific

Synonyme: 1-(1-(Phenylsulphonyl)-1H-pyrrol-3-yl)ethanone

OR10821-1G OR10821-5G
APOSOR10821-1GEA 0 EUR
APOSOR10821-1G APOSOR10821-5G
3-Acetyl-1-(phenylsulphonyl)pyrrole
3-Acetyl-1-(phenylsulphonyl)pyrrole
Formel: C₁₂H₁₁NO₃S
Molecular Weight: 249.284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067752
CAS-Nummer: 81453-98-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR