3-(Aminomethyl)-1H-indole oxalate

Lieferant: Apollo Scientific
Achtung

OR10823-500MG OR10823-50MG
APOSOR10823-500MGEA 145 EUR
APOSOR10823-500MG APOSOR10823-50MG
3-(Aminomethyl)-1H-indole oxalate
3-(Aminomethyl)-1H-indole oxalate
Formel: C₉H₁₀N₂·C₂H₂O₄
Molecular Weight: 236.224 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863232
CAS-Nummer: 296775-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR